Assay Details
| Gene Symbol: Zfp281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160251.1 | NP_001153723.1 | 1 - 2 | 153 | 98 | |
| GenBank mRNA | BC062153.1 | - | 1 - 2 | 103 | 6335572 | 98 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062153.1, NM_001160251.1 transcripts.
Set Membership: |
|

Zfp281
Genomic Map