Assay Details
| Gene Symbol: BC051142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001177.2 | NP_001001177.2 | 16 - 17 | 963 | 100 | |
| NM_001163855.1 | NP_001157327.1 | 20 - 21 | 736 | 76 | ||
| GenBank mRNA | AK077077.1 | - | 16 - 17 | 950 | 100 | |
| AK133031.1 | - | 15 - 16 | 796 | 100 | ||
| BC051142.1 | - | 20 - 21 | 736 | 6740688 | 76 | |
More Information
Set Membership: |
|

BC051142
Genomic Map