Assay Details
| Gene Symbol: Kbtbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024135.2 | NP_001019306.2 | 1 | 2114 | 82 | |
| GenBank mRNA | AK135854.1 | - | 2 | 1440 | 82 | |
| BC096019.1 | - | 1 | 2114 | 6830769 | 82 | |
More Information
Set Membership: |
|

Kbtbd7
Genomic Map