Assay Details
| Gene Symbol: Prrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025570.1 | NP_001020741.1 | 1 - 2 | 1254 | 92 | |
| NM_011127.2 | NP_035257.1 | 1 - 2 | 1254 | 92 | ||
| NM_175686.3 | NP_783617.2 | 1 - 2 | 1254 | 92 | ||
| GenBank mRNA | AB221686.1 | - | 1 - 2 | 239 | 92 | |
| AK019503.1 | - | 1 - 2 | 65 | 92 | ||
| AK031075.1 | - | 1 - 2 | 220 | 92 | ||
| AK032773.1 | - | 1 - 2 | 821 | 92 | ||
| AK084387.1 | - | 1 - 2 | 1272 | 92 | ||
| AK089006.1 | - | 1 - 2 | 1086 | 92 | ||
| BC092372.1 | - | 1 - 2 | 603 | 30533016 | 92 | |
| L06502.1 | - | 1 - 2 | 419 | 92 | ||
| U03873.1 | - | 1 - 2 | 1260 | 92 | ||
| X59725.1 | - | 1 - 2 | 380 | 92 | ||
More Information
Set Membership: |
|

Prrx1
Genomic Map