Assay Details
| Gene Symbol: Gm9513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128510.1 | NP_001121982.1 | 2 - 3 | 171 | 110 | |
| GenBank mRNA | EU703629.1 | - | 2 - 3 | 171 | 110 | |
More Information
Set Membership: |
|

Gm9513
Genomic Map