Assay Details
| Gene Symbol: Rpl35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130484.1 | NP_001123956.1 | 2 - 3 | 118 | 106 | |
| NM_021338.3 | NP_067313.2 | 2 - 3 | 148 | 168 | ||
| GenBank mRNA | AV486753.1 | - | 2 - 3 | 154 | 168 | |
| CJ043213.1 | - | 2 - 3 | 119 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130484.1, NM_021338.3 transcripts.
Set Membership: |
|

Rpl35a
Genomic Map