Assay Details
Gene Symbol: Zfp33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CA546228.1 | - | 2 - 3 | 183 | 129 | |
CN678741.1 | - | 2 - 3 | 164 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
