Assay Details
| Gene Symbol: Rpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025592.3 | NP_079868.1 | 3 - 4 | 262 | 125 | |
| GenBank mRNA | AK010639.2 | - | 3 - 4 | 263 | 125 | |
| AK012335.1 | - | 3 - 4 | 263 | 125 | ||
| AK151707.1 | - | 3 - 4 | 257 | 125 | ||
| AK169650.1 | - | 3 - 4 | 263 | 125 | ||
| BC025884.1 | - | 3 - 4 | 244 | 125 | ||
| BC094634.1 | - | 3 - 4 | 246 | 1333082 | 125 | |
| BC125652.1 | - | 3 - 4 | 244 | 40130167 | 125 | |
| BC125656.1 | - | 3 - 4 | 244 | 40130171 | 125 | |
More Information
Set Membership: |
|

Rpl35
Genomic Map