Assay Details
| Gene Symbol: Rpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026055.1 | NP_080331.1 | 1 - 2 | 58 | 107 | |
| GenBank mRNA | AK002591.1 | - | 1 - 2 | 56 | 107 | |
| AK005511.1 | - | 1 - 2 | 58 | 107 | ||
| AK007821.1 | - | 1 - 2 | 60 | 107 | ||
| AK008853.1 | - | 1 - 2 | 56 | 107 | ||
| AK013381.1 | - | 1 - 2 | 62 | 107 | ||
| AK028139.1 | - | 1 - 2 | 58 | 107 | ||
| AK088761.1 | - | 1 - 2 | 63 | 107 | ||
| AK146772.1 | - | 1 - 2 | 60 | 107 | ||
| AK168773.1 | - | 1 - 2 | 56 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002591.1, AK005511.1, AK007821.1, AK008853.1, AK013381.1, AK028139.1, AK088761.1, AK146772.1, AK168773.1, NM_026055.1 transcripts.
Set Membership: |
|

Rpl39
Genomic Map