Assay Details
| Gene Symbol: Nlrp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139925.1 | - | 2 - 3 | 355 | 107 | |
| AY364009.1 | - | 271 | 107 | |||
| AY596202.1 | - | 1 - 2 | 271 | 107 | ||
| DV651562.1 | - | 2 - 3 | 355 | 107 | ||
More Information
Set Membership: |
|

Nlrp9a
Genomic Map