Assay Details
| Gene Symbol: Nlrp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139925.1 | - | 9 - 10 | 2948 | 89 | |
| AK145355.1 | - | 8 - 9 | 2624 | 89 | ||
| AY364009.1 | - | 2527 | 89 | |||
| AY596202.1 | - | 7 - 8 | 2698 | 89 | ||
| DV652646.1 | - | 2 - 3 | 220 | 89 | ||
More Information
Set Membership: |
|

Nlrp9a
Genomic Map