Assay Details
| Gene Symbol: Hamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183257.3 | NP_899080.1 | 2 - 3 | 196 | 124 | |
| GenBank mRNA | AK007975.1 | - | 2 - 3 | 186 | 124 | |
| AK008993.1 | - | 2 - 3 | 197 | 124 | ||
| AY232841.1 | - | 2 - 3 | 185 | 124 | ||
| BC086895.1 | - | 2 - 3 | 193 | 6765293 | 124 | |
More Information
Set Membership: |
|

Hamp2
Genomic Map