Assay Details
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113187.1 | NP_001106658.1 | 3 | 714 | 90 | |
| GenBank mRNA | BG081161.2 | - | 3 | 440 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdc1b| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113187.1 | NP_001106658.1 | 3 | 714 | 90 | |
| GenBank mRNA | BG081161.2 | - | 3 | 440 | 90 | |
Set Membership: |
|