Assay Details
| Gene Symbol: Zfp33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009327.2 | - | 1 | 1395 | 105 | |
| AK049668.1 | - | 1740 | 105 | |||
| BC046593.1 | - | 1 | 1561 | 105 | ||
| BG073531.2 | - | 1 | 235 | 105 | ||
More Information
Set Membership: |
|

Zfp33b
Genomic Map