Assay Details
| Gene Symbol: Naip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010870.2 | NP_035000.2 | 15 | 4413 | 149 | |
| GenBank mRNA | AF135492.1 | - | 15 | 4399 | 149 | |
| AF135493.1 | - | 13 | 1971 | 149 | ||
| AF381771.1 | - | 15 | 4396 | 149 | ||
| AK037418.1 | - | 1 | 292 | 149 | ||
| AY146994.1 | - | 14 | 4229 | 149 | ||
| AY146995.1 | - | 14 | 4229 | 149 | ||
| AY146996.1 | - | 14 | 4260 | 149 | ||
| AY146997.1 | - | 14 | 4265 | 149 | ||
| AY146998.1 | - | 14 | 4260 | 149 | ||
| AY146999.1 | - | 14 | 4221 | 149 | ||
| BC070433.1 | - | 14 | 4227 | 6850660 | 149 | |
More Information
Set Membership: |
|

Naip5
Genomic Map