Assay Details
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025381.2 | NP_079657.1 | 2 | 224 | 102 | |
GenBank mRNA | AK003419.1 | - | 2 | 221 | 102 | |
AK007327.1 | - | 2 | 225 | 102 | ||
AK152153.1 | - | 2 | 221 | 102 | ||
AK152929.1 | - | 2 | 248 | 102 | ||
AK152959.1 | - | 2 | 221 | 102 | ||
BC016553.1 | - | 2 | 234 | 2812592 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
