Assay Details
| Gene Symbol: Ugt2b37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053215.3 | NP_444445.2 | 1 | 518 | 85 | |
| GenBank mRNA | AK002736.1 | - | 1 | 535 | 85 | |
| BC019487.1 | - | 1 | 518 | 4237718 | 85 | |
More Information
Set Membership: |
|

Ugt2b37
Genomic Map