Assay Details
| Gene Symbol: Pcdha12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138663.2 | NP_619604.1 | 1 | 1020 | 68 | |
| GenBank mRNA | AB008180.1 | - | 1 | 913 | 68 | |
| AY013759.1 | - | 1 | 1020 | 68 | ||
More Information
Set Membership: |
|

Pcdha12
Genomic Map