Assay Details
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153780.3 | NP_722475.1 | 5 | 1310 | 122 | |
| GenBank mRNA | AK011776.1 | - | 5 | 1287 | 122 | |
| AK138138.1 | - | 5 | 1311 | 122 | ||
| BC023890.1 | - | 5 | 1269 | 5252244 | 122 | |
More Information
Set Membership: |
|

2610044O15Rik8
Genomic Map