Assay Details
| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183160.3 | NP_898983.1 | 2 | 524 | 90 | |
| GenBank mRNA | AK086895.1 | - | 2 | 526 | 90 | |
| AK142952.1 | - | 2 | 528 | 90 | ||
| AK143762.1 | - | 2 | 526 | 90 | ||
| BC087877.1 | - | 2 | 527 | 30302311 | 90 | |
More Information
Set Membership: |
|

Tmem252
Genomic Map