Assay Details
| Gene Symbol: Cxcr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC031665.1 | - | 2 | 131 | 3592479 | 105 |
| BC098322.1 | - | 2 | 121 | 2864967 | 105 | |
| U59760.1 | - | 2 | 83 | 105 | ||
More Information
Set Membership: |
|

Cxcr4
Genomic Map