Assay Details
| Gene Symbol: Il6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031168.1 | NP_112445.1 | 2 - 3 | 233 | 75 | |
| GenBank mRNA | AK089780.1 | - | 2 - 3 | 252 | 75 | |
| AK150440.1 | - | 2 - 3 | 252 | 75 | ||
| AK152189.1 | - | 1 - 2 | 239 | 75 | ||
| BC132458.1 | - | 2 - 3 | 226 | 40130735 | 75 | |
| BC138766.1 | - | 2 - 3 | 226 | 8861788 | 75 | |
| BC145409.1 | - | 2 - 3 | 226 | 75 | ||
| DQ788722.1 | - | 2 - 3 | 202 | 75 | ||
| J03783.1 | - | 2 - 3 | 250 | 75 | ||
| X06203.1 | - | 2 - 3 | 235 | 75 | ||
| X54542.1 | - | 2 - 3 | 233 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
- Authors
- Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
- Journal
- J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 184 (2010)
- Product Usage
- -

Il6
Genomic Map