Assay Details
| Gene Symbol: Il12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128214.1 | - | 4 - 5 | 507 | 63 | |
| AF128215.1 | - | 4 - 5 | 507 | 63 | ||
| AK162981.1 | - | 4 - 5 | 531 | 63 | ||
| BC103608.1 | - | 4 - 5 | 513 | 40046200 | 63 | |
| BC103609.1 | - | 4 - 5 | 513 | 40046202 | 63 | |
| BC103610.1 | - | 4 - 5 | 513 | 40046204 | 63 | |
| BC103614.1 | - | 4 - 5 | 513 | 40046206 | 63 | |
| M86671.1 | - | 4 - 5 | 508 | 63 | ||
More Information
Set Membership: |
|

Il12b
Genomic Map