Assay Details
| Gene Symbol: LacZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_013364_LacZ.0 | - | 1305 | 77 | ||
| Gene Symbol: YFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY189981_YFP.0 | - | 77 | |||
More Information
Set Membership: |
|

LacZ
Genomic Map