Assay Details
| Gene Symbol: YFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY189981_YFP.0 | - | 1978 | 61 | ||
| Gene Symbol: YFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY189981_YFP.0 | - | 1417 | 61 | ||
| Gene Symbol: Enhanced GFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U55761.1 | - | 472 | 61 | ||
More Information
Set Membership: |
|

YFP
Genomic Map