Assay Details
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971614.1 | XP_027827415.1 | 18 - 19 | 1521 | 59 | |
XM_027971615.1 | XP_027827416.1 | 18 - 19 | 1521 | 59 | ||
XM_027971616.1 | XP_027827417.1 | 18 - 19 | 1521 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
