Assay Details
| Gene Symbol: FGF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012656.4 | XP_004012705.1 | 4 - 5 | 1882 | 73 | |
| XM_027974971.1 | XP_027830772.1 | 4 - 5 | 1948 | 73 | ||
| XM_027974973.1 | XP_027830774.1 | 4 - 5 | 1798 | 73 | ||
More Information
Set Membership: |
|

FGF11
Genomic Map