Assay Details
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104810.2 | XP_014960296.1 | 2 - 3 | 461 | 67 | |
XM_015104811.2 | XP_014960297.1 | 2 - 3 | 461 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
