Assay Details
Gene Symbol: GSK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129740.1 | NP_001123212.1 | 9 - 10 | 1093 | 66 | |
XM_012178649.3 | XP_012034039.1 | 10 - 11 | 2162 | 66 | ||
XM_012179101.3 | XP_012034491.1 | 10 - 11 | 1570 | 66 | ||
XM_012180332.3 | XP_012035722.1 | 10 - 11 | 1919 | 66 | ||
XM_012181067.3 | XP_012036457.1 | 10 - 11 | 1569 | 66 | ||
XM_027973494.1 | XP_027829295.1 | 10 - 11 | 1570 | 66 | ||
XM_027975301.1 | XP_027831102.1 | 9 - 10 | 1527 | 66 | ||
GenBank mRNA | EU239382.1 | - | 9 - 10 | 1093 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
