Assay Details
| Gene Symbol: OAZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290006.2 | NP_001276935.1 | 2 - 3 | 348 | 114 | |
| GenBank mRNA | GT644620.1 | - | 2 - 3 | 372 | 114 | |
| GT873426.1 | - | 2 - 3 | 267 | 114 | ||
More Information
Set Membership: |
|

OAZ2
Genomic Map