Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010780.4 | XP_004010829.1 | 4 - 5 | 833 | 86 | |
| XM_004010781.4 | XP_004010830.1 | 4 - 5 | 690 | 86 | ||
| XM_012182025.2 | XP_012037415.1 | 4 - 5 | 574 | 86 | ||
More Information
Set Membership: |
|

ZNF410
Genomic Map