Assay Details
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011781.3 | XP_004011830.1 | 4 - 5 | 367 | 101 | |
XM_004011782.4 | XP_004011831.1 | 4 - 5 | 309 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
