Assay Details
| Gene Symbol: SLC25A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142895.1 | NP_001136367.1 | 6 - 7 | 929 | 65 | |
| XM_027965725.1 | XP_027821526.1 | 6 - 7 | 947 | 65 | ||
| XM_027965726.1 | XP_027821527.1 | 5 - 6 | 1114 | 65 | ||
| GenBank mRNA | FJ422547.1 | - | 6 - 7 | 929 | 65 | |
More Information
Set Membership: |
|

SLC25A3
Genomic Map