Assay Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006589.3 | XP_004006638.1 | 4 - 5 | 429 | 69 | |
| XM_004006590.3 | XP_004006639.1 | 4 - 5 | 429 | 69 | ||
| XM_027967603.1 | XP_027823404.1 | 3 - 4 | 371 | 69 | ||
| XM_027967604.1 | XP_027823405.1 | 3 - 4 | 371 | 69 | ||
| GenBank mRNA | EU370541.1 | - | 2 - 3 | 326 | 69 | |
More Information
Set Membership: |
|

MYL6
Genomic Map