Assay Details
| Gene Symbol: TMEM79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002610.3 | XP_004002659.2 | 3 - 4 | 1147 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM79| Gene Symbol: TMEM79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002610.3 | XP_004002659.2 | 3 - 4 | 1147 | 64 | |
Set Membership: |
|