Assay Details
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142509.1 | NP_001135981.1 | 4 - 5 | 610 | 57 | |
XM_012158800.3 | XP_012014190.2 | 4 - 5 | 717 | 57 | ||
GenBank mRNA | FJ422550.1 | - | 4 - 5 | 610 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
