Assay Details
| Gene Symbol: ARAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165890.1 | NP_001159362.1 | 10 - 11 | 1253 | 56 | |
| XM_012106111.3 | XP_011961501.2 | 11 - 12 | 1410 | 56 | ||
| XM_012106112.3 | XP_011961502.2 | 11 - 12 | 1395 | 56 | ||
| GenBank mRNA | FJ937964.1 | - | 10 - 11 | 1253 | 56 | |
More Information
Set Membership: |
|

ARAF
Genomic Map