Assay Details
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009469.1 | NP_001009469.1 | 1 - 2 | 117 | 66 | |
XM_027973955.1 | XP_027829756.1 | 2 - 3 | 231 | 66 | ||
XM_027973956.1 | XP_027829757.1 | 2 - 3 | 316 | 66 | ||
XM_027973957.1 | XP_027829758.1 | 2 - 3 | 157 | 66 | ||
GenBank mRNA | X80024.1 | - | 117 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
