Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205153.1 | NP_001192082.1 | 6 - 7 | 728 | 60 | |
| XM_027976353.1 | XP_027832154.1 | 6 - 7 | 858 | 60 | ||
| XM_027976354.1 | XP_027832155.1 | 6 - 7 | 858 | 60 | ||
| GenBank mRNA | HQ263380.1 | - | 6 - 7 | 728 | 60 | |
More Information
Set Membership: |
|

HNF4A
Genomic Map