Assay Details
| Gene Symbol: BSCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102042.2 | XP_011957432.1 | 5 - 6 | 845 | 57 | |
| XM_012102044.3 | XP_011957434.1 | 5 - 6 | 860 | 57 | ||
| XM_012102045.3 | XP_011957435.1 | 5 - 6 | 879 | 57 | ||
| XM_012102047.2 | XP_011957437.1 | 5 - 6 | 1214 | 57 | ||
| XM_012102048.3 | XP_011957438.1 | 4 - 5 | 613 | 57 | ||
| XM_012102049.3 | XP_011957439.1 | 4 - 5 | 705 | 57 | ||
| XM_027960097.1 | XP_027815898.1 | 5 - 6 | 843 | 57 | ||
| XM_027960098.1 | XP_027815899.1 | 5 - 6 | 843 | 57 | ||
| XM_027960099.1 | XP_027815900.1 | 5 - 6 | 843 | 57 | ||
More Information
Set Membership: |
|

BSCL2
Genomic Map