Assay Details
| Gene Symbol: PRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978633.1 | XP_027834434.1 | 7 - 8 | 1039 | 60 | |
| XM_027978634.1 | XP_027834435.1 | 7 - 8 | 637 | 60 | ||
| XM_027978635.1 | XP_027834436.1 | 6 - 7 | 673 | 60 | ||
| XM_027978636.1 | XP_027834437.1 | 8 - 9 | 774 | 60 | ||
More Information
Set Membership: |
|

PRMT1
Genomic Map