Assay Details
| Gene Symbol: FBXL21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129738.1 | NP_001123210.1 | 1 - 2 | 360 | 104 | |
| XM_027969162.1 | XP_027824963.1 | 1 - 2 | 1472 | 104 | ||
| GenBank mRNA | EU239380.1 | - | 1 - 2 | 360 | 104 | |
| EU239381.1 | - | 1 - 2 | 360 | 104 | ||
More Information
Set Membership: |
|

FBXL21P
Genomic Map