Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969496.1 | XP_027825297.1 | 7 - 8 | 981 | 60 | |
XM_027969497.1 | XP_027825298.1 | 7 - 8 | 973 | 60 | ||
XM_027969498.1 | XP_027825299.1 | 7 - 8 | 2822 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
