Assay Details
| Gene Symbol: ADAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011387.3 | XP_004011436.1 | 1 - 2 | 151 | 76 | |
| XM_012183021.3 | XP_012038411.1 | 1 - 2 | 151 | 76 | ||
| XM_027972505.1 | XP_027828306.1 | 1 - 2 | 151 | 76 | ||
| XM_027972506.1 | XP_027828307.1 | 1 - 2 | 151 | 76 | ||
More Information
Set Membership: |
|

ADAT2
Genomic Map