Assay Details
| Gene Symbol: LOC101108520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017934.4 | XP_004017983.1 | 7 - 8 | 1188 | 78 | |
| XM_004017935.4 | XP_004017984.1 | 7 - 8 | 1150 | 78 | ||
| XM_012098909.3 | XP_011954299.1 | 7 - 8 | 1148 | 78 | ||
| XM_012098910.3 | XP_011954300.1 | 7 - 8 | 1019 | 78 | ||
More Information
Set Membership: |
|

LOC101108520
Genomic Map