Assay Details
| Gene Symbol: KRT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974574.1 | XP_027830375.1 | 5 - 6 | 1720 | 63 | |
| XM_027974575.1 | XP_027830376.1 | 5 - 6 | 1733 | 63 | ||
More Information
Set Membership: |
|

KRT10
Genomic Map