Assay Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978277.1 | XP_027834078.1 | 3 - 4 | 345 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS16| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978277.1 | XP_027834078.1 | 3 - 4 | 345 | 87 | |
Set Membership: |
|