Assay Details
| Gene Symbol: KXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969395.1 | XP_027825196.1 | 3 - 4 | 375 | 58 | |
| XM_027969396.1 | XP_027825197.1 | 3 - 4 | 447 | 58 | ||
More Information
Set Membership: |
|

KXD1
Genomic Map