Assay Details
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006923.4 | XP_004006972.1 | 5 - 6 | 873 | 62 | |
XM_012175501.3 | XP_012030891.1 | 5 - 6 | 801 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
