Assay Details
Gene Symbol: RNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016160.4 | XP_004016209.1 | 3 - 4 | 625 | 64 | |
XM_027979397.1 | XP_027835198.1 | 3 - 4 | 410 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
